Methyl-3-bromo-1H-indazole-5-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 3-bromo-1H-indazole-5-carboxylate

068463-1G 068463-5G
101936-258EA 223.08 USD
101936-258 101936-260
Methyl-3-bromo-1H-indazole-5-carboxylate ≥95%
Methyl 3-bromo-1H-indazole-5-carboxylate

Hazard: Irritant

Formula: C₉H₇BrN₂O₂
MW: 255.0681 g/mol
MDL Number: MFCD11045401
CAS Number: 1086391-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR