Methyl-3-iodo-1H-indazole-5-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 3-iodo-1H-indazole-5-carboxylate
068462-1G
068462-5G
101936-254EA
815.38
USD
101936-254
101936-256
Methyl-3-iodo-1H-indazole-5-carboxylate ≥95%
Methyl 3-iodo-1H-indazole-5-carboxylate
Hazard: Irritant
Formula:
C₉H₇IN₂O₂ MW: 302.0700 g/mol |
MDL Number:
MFCD06739144 CAS Number: 885271-25-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...