3-Methylamino-5-bromo-1H-pyrazolo[3,4-b]pyridine ≥95%

Supplier: Matrix Scientific

068461-250MG 068461-500MG 068461-1G
101936-248EA 1309.23 USD
101936-248 101936-250 101936-252
3-Methylamino-5-bromo-1H-pyrazolo[3,4-b]pyridine ≥95%
3-Methylamino-5-bromo-1H-pyrazolo[3,4-b]pyridine

Hazard: Irritant

Formula: C₈H₈BrN₃
MDL Number: MFCD18432771
CAS Number: 1289189-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR