1-Methyl-1H-indazole-5-boronic acid pinacol ester ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Methylindazole-5-boronic acid pinacol ester, 1-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole

068459-250MG 068459-500MG 068459-1G
101936-242EA 1221.54 USD
101936-242 101936-244 101936-246
1-Methyl-1H-indazole-5-boronic acid pinacol ester ≥95%
1-Methyl-1H-indazole-5-boronic acid pinacol ester

Hazard: Irritant

Formula: C₁₄H₁₉BN₂O₂
MW: 258.123 g/mol
MDL Number: MFCD15071438
CAS Number: 1235469-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR