tert-Butyl-5-bromo-3-(chloromethyl)pyrrolo[2,3-b]pyridine-1-carboxylate ≥95%

Supplier: Matrix Scientific

068458-250MG 068458-500MG 068458-1G
101936-236EA 801.54 USD
101936-236 101936-238 101936-240
tert-Butyl-5-bromo-3-(chloromethyl)pyrrolo[2,3-b]pyridine-1-carboxylate ≥95%
tert-Butyl-5-bromo-3-(chloromethyl)pyrrolo[2,3-b]pyridine-1-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₄BrClN₂O₂
MDL Number: MFCD17015935
CAS Number: 1234616-42-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR