Methyl-3-bromo-7-azaindole-4-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 3-bromo-1H-pyrrolo[2,3-b]pyridine-4-carboxylate

068457-250MG 068457-500MG 068457-1G
101936-230EA 1018.46 USD
101936-230 101936-232 101936-234
Methyl-3-bromo-7-azaindole-4-carboxylate ≥95%
Methyl-3-bromo-7-azaindole-4-carboxylate

Hazard: Irritant

Formula: C₉H₇BrN₂O₂
MW: 255.08 g/mol
MDL Number: MFCD12962695
CAS Number: 1190310-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR