3-Bromo-7-azaindole-5-carboxylic acid methyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 3-bromo-1H-pyrrolo[2,3-b]pyridine-5-carboxylate

068456-250MG 068456-500MG
101936-226EA 640 USD
101936-226 101936-228
3-Bromo-7-azaindole-5-carboxylic acid methyl ester ≥95%
3-Bromo-7-azaindole-5-carboxylic acid methyl ester

Hazard: Irritant

Formula: C₉H₇BrN₂O₂
MW: 255.08 g/mol
MDL Number: MFCD12962903
CAS Number: 1190322-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR