5-Carboxyoxindole ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Oxoindoline-5-carboxylic Acid, 2-Oxindole-5-carboxylic acid
068455-1G
068455-5G
101936-222EA
104.62
USD
101936-222
101936-224
5-Carboxyoxindole ≥95%
5-Carboxyoxindole
Hazard: Irritant
Formula:
C₉H₇NO₃ MW: 177.1600 g/mol |
MDL Number:
MFCD03411961 CAS Number: 102359-00-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...