4-Chloropyridine-2,3-diamine ≥95%

Supplier: Matrix Scientific

Synonyms: 2,3-Diamino-4-chloropyridine

068451-250MG 068451-500MG
101936-204EA 495.38 USD
101936-204 101936-206
4-Chloropyridine-2,3-diamine ≥95%
4-Chloropyridine-2,3-diamine

Hazard: Irritant

Formula: C₅H₆ClN₃
MW: 143.57 g/mol
MDL Number: MFCD13181777
CAS Number: 24484-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR