7-Chloroimidazo[1,2-a]pyridine ≥95%

Supplier: Matrix Scientific

068450-1G 068450-5G
101936-200EA 41.54 USD
101936-200 101936-202
7-Chloroimidazo[1,2-a]pyridine ≥95%
7-Chloroimidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₇H₅ClN₂
MW: 152.5800 g/mol
MDL Number: MFCD08275126
CAS Number: 4532-25-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR