3-(Methylsulfonyl)azetidine hydrochloride ≥95%
Supplier: Matrix Scientific
068448-250MG
068448-500MG
068448-1G
101936-188EA
246.15
USD
101936-188
101936-190
101936-192
3-(Methylsulfonyl)azetidine hydrochloride ≥95%
3-(Methylsulfonyl)azetidine hydrochloride
Hazard: Irritant
Formula:
C4H10ClNO2S |
MDL Number:
MFCD22416367 CAS Number: 1400764-60-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...