3-Hydroxy-1-methylazetidine hydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
1-Methyl-3-azetidinol Hydrochloride, 1-Methylazetidin-3-ol hydrochloride
068447-250MG
068447-500MG
068447-1G
101936-182EA
172.31
USD
101936-182
101936-184
101936-186
3-Hydroxy-1-methylazetidine hydrochloride ≥95%
3-Hydroxy-1-methylazetidine hydrochloride
Hazard: Irritant
Formula:
C₄H₉NO·ClH MW: 123.581 g/mol |
MDL Number:
MFCD16605978 CAS Number: 26687-49-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...