tert-Butyl-N-[(1S,5R)-3-azabicyclo[3.1.0]hexan-6-yl]carbamate ≥95%

Supplier: Matrix Scientific

068444-100MG 068444-250MG 068444-500MG
101936-166EA 306.15 USD
101936-166 101936-168 101936-170
tert-Butyl-N-[(1S,5R)-3-azabicyclo[3.1.0]hexan-6-yl]carbamate ≥95%
tert-Butyl-N-[(1S,5R)-3-azabicyclo[3.1.0]hexan-6-yl]carbamate

Hazard: Irritant

Formula: C₁₀H₁₈N₂O₂
MDL Number: MFCD12198677
CAS Number: 134575-17-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR