2-Trifluoromethoxybenzylhydrazine dihydrochloride ≥95%

Supplier: Matrix Scientific

068441-250MG 068441-500MG
101936-152EA 378.46 USD
101936-152 101936-154
2-Trifluoromethoxybenzylhydrazine dihydrochloride ≥95%
2-Trifluoromethoxybenzylhydrazine dihydrochloride

Hazard: Irritant

Formula: C₈H₁₁Cl₂F₃N₂O
MDL Number: MFCD18425619
CAS Number: 887595-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR