3-(Hydrazinomethyl)pyridine dihydrochloride ≥95%

Supplier: Matrix Scientific

068439-250MG 068439-500MG
101936-142EA 436.92 USD
101936-142 101936-144
3-(Hydrazinomethyl)pyridine dihydrochloride ≥95%
3-(Hydrazinomethyl)pyridine dihydrochloride

Hazard: Irritant

Formula: C₆H₁₁Cl₂N₃
MDL Number: MFCD10686872
CAS Number: 6978-97-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR