(2-Fluorobenzyl)hydrazine ≥95%
Supplier: Matrix Scientific
068434-1G
068434-5G
101936-122EA
81.54
USD
101936-122
101936-124
(2-Fluorobenzyl)hydrazine ≥95%
(2-Fluorobenzyl)hydrazine
Hazard: Irritant
Formula:
C₇H₉FN₂ MW: 140.1582 g/mol |
MDL Number:
MFCD07785903 CAS Number: 51859-98-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...