3-Fluorobenzylhydrazine ≥95%

Supplier: Matrix Scientific

068433-1G 068433-5G
101936-118EA 346.15 USD
101936-118 101936-120
3-Fluorobenzylhydrazine ≥95%
3-Fluorobenzylhydrazine

Hazard: Irritant

Formula: C₇H₉FN₂
MDL Number: MFCD07785902
CAS Number: 51421-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR