4-Fluorobenzylhydrazine ≥95%

Supplier: Matrix Scientific

068432-1G 068432-5G
101936-114EA 640 USD
101936-114 101936-116
4-Fluorobenzylhydrazine ≥95%
4-Fluorobenzylhydrazine

Hazard: Irritant

Formula: C₇H₉FN₂
MDL Number: MFCD06797341
CAS Number: 699-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR