2-Bromo-5H-pyrrolo[2,3-b]pyrazine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-4,7-diazaindole

068431-1G 068431-5G 068431-250MG
101936-110EA 101.54 USD
101936-110 101936-112 102663-834
2-Bromo-5H-pyrrolo[2,3-b]pyrazine ≥95%
2-Bromo-5H-pyrrolo[2,3-b]pyrazine

Hazard: Irritant

Formula: C₆H₄BrN₃
MW: 198.0201 g/mol
MDL Number: MFCD09834822
CAS Number: 875781-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR