2-Pyridinesulfonamide ≥95%

Supplier: Matrix Scientific

Synonyms: Pyridine-2-sulphonamide

068430-250MG 068430-500MG 068430-1G
101936-104EA 133.85 USD
101936-104 101936-106 101936-108
2-Pyridinesulfonamide ≥95%
2-Pyridinesulfonamide

Hazard: Irritant

Formula: C₅H₆N₂O₂S
MW: 158.18 g/mol
MDL Number: MFCD09260336
CAS Number: 63636-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR