4-Pyridinesulfonamide ≥95%

Supplier: Matrix Scientific

068429-250MG 068429-500MG 068429-1G
101936-098EA 200 USD
101936-098 101936-100 101936-102
4-Pyridinesulfonamide ≥95%
4-Pyridinesulfonamide

Hazard: Irritant

Formula: C₅H₆N₂O₂S
MDL Number: MFCD12913636
CAS Number: 65938-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR