5-Chloro-3H-imidazo[4,5-b]pyridine ≥95%

Supplier: Matrix Scientific

068428-1G 068428-5G
101936-094EA 1396.92 USD
101936-094 101936-096
5-Chloro-3H-imidazo[4,5-b]pyridine ≥95%
5-Chloro-3H-imidazo[4,5-b]pyridine

Hazard: Irritant

Formula: C₆H₄ClN₃
MW: 153.5700 g/mol
MDL Number: MFCD01646134
CAS Number: 52090-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR