4-Chloro-5,7-dihydro-6H-pyrrolo[2,3-d]pyrimidin-6-one ≥95%

Supplier: Matrix Scientific

Synonyms: 4-chloro-5H,6H,7H-pyrrolo[2,3-d]pyrimidin-6-one, 4-Chloro-5H-pyrrolo[2,3-d]pyrimidin-6(7H)-one

068424-1G 068424-5G
101936-074EA 346.15 USD
101936-074 101936-076
4-Chloro-5,7-dihydro-6H-pyrrolo[2,3-d]pyrimidin-6-one ≥95%
4-Chloro-5,7-dihydro-6H-pyrrolo[2,3-d]pyrimidin-6-one

Hazard: Irritant

Formula: C₆H₄ClN₃O
MW: 169.57 g/mol
MDL Number: MFCD09832903
CAS Number: 346599-63-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR