1,3-Dihydro-2H-pyrrolo[3,2-c]pyridin-2-one ≥95%

Supplier: Matrix Scientific

068423-250MG 068423-500MG
101936-070EA 727.69 USD
101936-070 101936-072
1,3-Dihydro-2H-pyrrolo[3,2-c]pyridin-2-one ≥95%
1,3-Dihydro-2H-pyrrolo[3,2-c]pyridin-2-one

Hazard: Irritant

Formula: C₇H₆N₂O
MDL Number: MFCD09832902
CAS Number: 134682-54-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR