4-Methoxypyrimidin-2-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-4-methoxypyrimidine

068422-1G 068422-5G
101936-066EA 150.77 USD
101936-066 101936-068
4-Methoxypyrimidin-2-amine ≥95%
4-Methoxypyrimidin-2-amine

Hazard: Irritant

Formula: C₅H₇N₃O
MW: 125.13 g/mol
MDL Number: MFCD00619233
CAS Number: 155-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR