2-Chloro-9H-purine ≥95%

Supplier: Matrix Scientific

068421-1G 068421-5G
101936-062EA 323.08 USD
101936-062 101936-064
2-Chloro-9H-purine ≥95%
2-Chloro-9H-purine

Hazard: Irritant/Refrigerate

Formula: C₅H₃ClN₄
MW: 154.5600 g/mol
MDL Number: MFCD00137826
CAS Number: 1681-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR