5-Bromo-1H-pyrazolo[3,4-b]pyridin-3-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Amino-5-bromo-1H-pyrazolo[3,4-b]pyridine

068420-1G 068420-5G
101936-058EA 720 USD
101936-058 101936-060
5-Bromo-1H-pyrazolo[3,4-b]pyridin-3-amine ≥95%
5-Bromo-1H-pyrazolo[3,4-b]pyridin-3-amine

Hazard: Irritant

Formula: C₆H₅BrN₄
MW: 213.0400 g/mol
MDL Number: MFCD08059261
CAS Number: 405224-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR