tert-Butyl-2-methyl-4-oxopiperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Methylpiperidin-4-one, N-BOC protected, tert-Butyl 2-methyl-4-oxopiperidine-1-carboxylate

068413-1G 068413-5G
101936-026EA 346.15 USD
101936-026 101936-028
tert-Butyl-2-methyl-4-oxopiperidine-1-carboxylate ≥95%
tert-Butyl-2-methyl-4-oxopiperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₉NO₃
MW: 213.2735 g/mol
MDL Number: MFCD04035608
CAS Number: 190906-92-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR