2-(1-tert-Butoxycarbonylazetidin-3-yl)acetic acid ≥95%

Supplier: Matrix Scientific

068411-250MG 068411-500MG 068411-1G
101936-014EA 233.85 USD
101936-014 101936-016 101936-018
2-(1-tert-Butoxycarbonylazetidin-3-yl)acetic acid ≥95%
2-(1-tert-Butoxycarbonylazetidin-3-yl)acetic acid

Hazard: Irritant

Formula: C₁₀H₁₇NO₄
MDL Number: MFCD02179028
CAS Number: 183062-96-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR