1H-Pyrazolo[3,4-b]pyridine-5-carboxylic acid ≥95%

Supplier: Matrix Scientific

068409-250MG 068409-500MG
101936-006EA 215.38 USD
101936-006 101936-008
1H-Pyrazolo[3,4-b]pyridine-5-carboxylic acid ≥95%
1H-Pyrazolo[3,4-b]pyridine-5-carboxylic acid

Hazard: Irritant

Formula: C₇H₅N₃O₂
MDL Number: MFCD09832891
CAS Number: 952182-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR