1H-Pyrrolo[2,3-b]pyridine-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 7-Azaindole-2-carboxylic acid

068408-1G 068408-5G
101936-002EA 756.92 USD
101936-002 101936-004
1H-Pyrrolo[2,3-b]pyridine-2-carboxylic acid ≥95%
1H-Pyrrolo[2,3-b]pyridine-2-carboxylic acid

Hazard: Irritant

Formula: C₈H₆N₂O₂
MW: 162.1500 g/mol
MDL Number: MFCD06659752
CAS Number: 136818-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR