4-Amino-2-piperidinone trifluoroacetate ≥95%

Supplier: Matrix Scientific

068407-500MG 068407-1G
101935-998EA 815.38 USD
101935-998 101936-000
4-Amino-2-piperidinone trifluoroacetate ≥95%
4-Amino-2-piperidinone trifluoroacetate

Hazard: Irritant

Formula: C₇H₁₁F₃N₂O₃
MDL Number: MFCD17016140
CAS Number: 5513-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR