4-Amino-1-methylpyridin-2(1H)-one ≥95%

Supplier: Matrix Scientific

068406-1G 068406-5G
101935-994EA 1338.46 USD
101935-994 101935-996
4-Amino-1-methylpyridin-2(1H)-one ≥95%
4-Amino-1-methylpyridin-2(1H)-one

Hazard: Irritant

Formula: C₆H₈N₂O
MW: 124.14 g/mol
MDL Number: MFCD09832890
CAS Number: 952182-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR