4-Amino-2-hydroxypyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Amino-2-pyridone, 4-Aminopyridin-2(1H)-one, 4-Amino-2(1H)-pyridinone, 4-Amino-2-pyridinol, 4-Amino-pyridin-2-ol, 4-amino-1,2-dihydropyridin-2-one, 3-Deazacytosine, 4-Amino-1H-pyridin-2-one

068405-1G 068405-5G
101935-990EA 113.85 USD
101935-990 101935-992
4-Amino-2-hydroxypyridine ≥95%
4-Amino-2-hydroxypyridine

Hazard: Irritant

Formula: C₅H₆N₂O
MW: 110.12 g/mol
MDL Number: MFCD02682913
CAS Number: 38767-72-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR