Methyl-4-chloro-7-methoxyquinoline-6-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 4-chloro-7-methoxyquinoline-6-carboxylate
068403-250MG
068403-500MG
101935-980EA
756.92
USD
101935-980
101935-982
Methyl-4-chloro-7-methoxyquinoline-6-carboxylate ≥95%
Methyl 4-chloro-7-methoxyquinoline-6-carboxylate
Hazard: Irritant
Formula:
C₁₂H₁₀ClNO₃ MW: 251.6700 g/mol |
MDL Number:
MFCD17016133 CAS Number: 205448-66-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...