4-(2H-Tetrazol-5-yl)benzenesulfonyl chloride ≥95%

Supplier: Matrix Scientific

068401-250MG 068401-500MG
101935-972EA 520 USD
101935-972 101935-974
4-(2H-Tetrazol-5-yl)benzenesulfonyl chloride ≥95%
4-(2H-Tetrazol-5-yl)benzenesulfonyl chloride

Hazard: Irritant

Formula: C₇H₅ClN₄O₂S
MDL Number: MFCD18432765
CAS Number: 924964-20-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR