7-Bromoquinazoline ≥95%

Supplier: Matrix Scientific

068398-250MG 068398-500MG 068398-1G
101935-958EA 756.92 USD
101935-958 101935-960 101935-962
7-Bromoquinazoline ≥95%
7-Bromoquinazoline

Hazard: Irritant

Formula: C₈H₅BrN₂
MDL Number: MFCD07778500
CAS Number: 89892-22-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR