6-Bromoquinazoline ≥95%

Supplier: Matrix Scientific

068397-1G 068397-5G
101935-956EA 136.92 USD
101935-956 102833-426
6-Bromoquinazoline ≥95%
6-Bromoquinazoline

Hazard: Irritant

Formula: C₈H₅BrN₂
MW: 209.0400 g/mol
MDL Number: MFCD08236695
CAS Number: 89892-21-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR