(±)-Boc-α-phosphonoglycine trimethyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-((tert-butoxycarbonyl)amino)-2-(dimethoxyphosphoryl)acetate

068395-5G 068395-25G
101935-946EA 306.15 USD
101935-946 102551-704
(±)-Boc-α-phosphonoglycine trimethyl ester ≥95%
(±)-Boc-α-phosphonoglycine trimethyl ester

Hazard: Irritant

Formula: C₁₀H₂₀NO₇P
Melting Pt: 67…71 °C
MDL Number: MFCD02178893
CAS Number: 89524-98-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR