4-Aminomethylpyrimidine dihydrochloride ≥95%
Supplier: Matrix Scientific
068394-250MG
068394-500MG
101935-940EA
930.77
USD
101935-940
101935-942
4-Aminomethylpyrimidine dihydrochloride ≥95%
4-Aminomethylpyrimidine dihydrochloride
Hazard: Irritant
Formula:
C₅H₉Cl₂N₃ MW: 182.05 g/mol |
MDL Number:
MFCD07781107 CAS Number: 618446-08-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...