2-Aminomethylpyrimidine ≥95%

Supplier: Matrix Scientific

Synonyms: Pyrimidin-2-ylmethanamine

068392-1G 068392-5G
101935-932EA 230.77 USD
101935-932 101935-934
2-Aminomethylpyrimidine ≥95%
2-Aminomethylpyrimidine

Hazard: Irritant

Formula: C₅H₇N₃
MW: 109.13 g/mol
MDL Number: MFCD06212857
CAS Number: 75985-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR