2-(Aminomethyl)pyrazine ≥95%

Supplier: Matrix Scientific

068391-1G 068391-5G
101935-928EA 233.85 USD
101935-928 101935-930
2-(Aminomethyl)pyrazine ≥95%
2-(Aminomethyl)pyrazine

Hazard: Irritant

Formula: C₅H₇N₃
MDL Number: MFCD00673149
CAS Number: 20010-99-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR