4-Aminomethylpyridazine hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Pyridazinemethanamine hydrochloride

068389-250MG 068389-500MG
101935-918EA 573.85 USD
101935-918 101935-920
4-Aminomethylpyridazine hydrochloride ≥95%
4-Aminomethylpyridazine hydrochloride

Hazard: Irritant

Formula: C₅H₈ClN₃
MDL Number: MFCD16988465
CAS Number: 1351479-13-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR