6-Hydroxy-4-methylpyridin-2(1H)-one ≥95%

Supplier: Matrix Scientific

068386-1G 068386-5G
101935-904EA 349.23 USD
101935-904 101935-906
6-Hydroxy-4-methylpyridin-2(1H)-one ≥95%
6-Hydroxy-4-methylpyridin-2(1H)-one

Hazard: Irritant

Formula: C₆H₇NO₂
MDL Number: MFCD17016132
CAS Number: 4664-16-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR