4-Amino-3,5-dimethylbenzonitrile ≥95%
Supplier: Matrix Scientific
Synonyms:
4-amino-3,5-dimethylbenzonitrile
068384-1G
068384-5G
101935-896EA
640
USD
101935-896
101935-898
4-Amino-3,5-dimethylbenzonitrile ≥95%
4-Amino-3,5-dimethylbenzonitrile
Hazard: Irritant
Formula:
C₉H₁₀N₂ MW: 146.1891 g/mol |
MDL Number:
MFCD07369841 CAS Number: 74896-24-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...