6-Amino-2,3-dihydro-2-methyl-1H-isoindol-1-one ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Amino-2-methylisoindolin-1-one

068383-250MG 068383-500MG
101935-892EA 815.38 USD
101935-892 101935-894
6-Amino-2,3-dihydro-2-methyl-1H-isoindol-1-one ≥95%
6-Amino-2,3-dihydro-2-methyl-1H-isoindol-1-one

Hazard: Irritant

Formula: C₉H₁₀N₂O
MW: 162.19 g/mol
MDL Number: MFCD13178503
CAS Number: 69189-26-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR