4-Methylthiopiperidine hydrochloride ≥95%

Supplier: Matrix Scientific

068380-250MG 068380-500MG
101935-878EA 466.15 USD
101935-878 101935-880
4-Methylthiopiperidine hydrochloride ≥95%
4-Methylthiopiperidine hydrochloride

Hazard: Irritant

Formula: C₆H₁₄ClNS
MDL Number: MFCD13188535
CAS Number: 208245-70-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR