Methyl-6-bromo-3-methoxy-2-methylbenzoate

Supplier: Matrix Scientific

067191-500MG 067191-1G 067191-5G
101933-452EA 403.08 USD
101933-452 101933-454 101933-456
Methyl-6-bromo-3-methoxy-2-methylbenzoate
Methyl-6-bromo-3-methoxy-2-methylbenzoate

Hazard: Irritant

Formula: C₁₀H₁₁BrO₃
MDL Number: MFCD16660004
CAS Number: 55289-16-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR