3-Hydroxy-2,3-dihydrobenzothiophene-1,1- dione
Supplier: Matrix Scientific
067183-500MG
067183-1G
101933-424EA
403.08
USD
101933-424
101933-426
3-Hydroxy-2,3-dihydrobenzothiophene-1,1- dione
3-Hydroxy-2,3-dihydrobenzothiophene-1,1- dione
Hazard: Irritant
Formula:
C₈H₈O₃S |
MDL Number:
MFCD20921609 CAS Number: 340774-59-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...