4-Ethyl-1-phenyl-1H-1,2,3-triazole

Supplier: Matrix Scientific

067176-500MG 067176-1G
101933-402EA 403.08 USD
101933-402 101933-404
4-Ethyl-1-phenyl-1H-1,2,3-triazole
4-Ethyl-1-phenyl-1H-1,2,3-triazole

Hazard: Irritant

Formula: C₁₀H₁₁N₃
Boiling Pt: 169…170 °C (12 Torr)
Melting Pt: 19 °C
MDL Number: MFCD20921600
CAS Number: 861519-47-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR